Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Di-n-propylformamide, 96%

$93.31 - $93.31

Chemical Identifiers

Chemical Name or Material N,N-Di-n-propylformamide
CAS 6282-00-4
Molecular Formula HCON(CH2CH2CH3)2
Formula Weight 129.20g/mol
MDL Number MFCD00043518
Beilstein 635928
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0885406

 
alfa aesar™
L08854-06
5g Each for $93.31
Description & Specifications

Specifications

Quantity 5g
Boiling Point 100° to 101°C (18mmHg)
Quantity 5g
Flash Point 88°C (190°F)
Chemical Name or Material N,N-Di-n-propylformamide
CAS 6282-00-4
Assay 96%
Molecular Formula HCON(CH2CH2CH3)2
Formula Weight 129.20g/mol
MDL Number MFCD00043518
Beilstein 635928
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number LQ2625000
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.