Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Di-n-propylethylenediamine, 97%

Chemical Identifiers

Chemical Name or Material N,N'-Di-n-propylethylenediamine
CAS 14165-22-1
Molecular Formula C8H20N2
Formula Weight 144.26
MDL Number MFCD00015199
UN Number UN2735
Beilstein 1734879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0467106

 
alfa aesar™
L0467106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0467114

 
alfa aesar™
L0467114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.26
CAS 14165-22-1
Chemical Name or Material N,N'-Di-n-propylethylenediamine
Boiling Point 186° to 189°C
Assay 97%
Quantity 5g
MDL Number MFCD00015199
Beilstein 1734879
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Flash Point 69°C (156°F)
Molecular Formula C8H20N2
UN Number UN2735
Health Hazard 1 Danger

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.