Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Di-n-hexylacetamide, 98+%

$320.18 - $320.18

Chemical Identifiers

Chemical Name or Material N,N-Di-n-hexylacetamide
CAS 16423-51-1
Molecular Formula C14H29NO
Formula Weight 227.39
MDL Number MFCD00082658
Beilstein 1772656
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1080518

 
alfa aesar™
L1080518
50g Each for $320.18
Description & Specifications

Specifications

Formula Weight 227.39
Assay ≥98%
Quantity 50g
Flash Point 106°C (223°F)
Molecular Formula C14H29NO
CAS 16423-51-1
Beilstein 1772656
Chemical Name or Material N,N-Di-n-hexylacetamide
Boiling Point 130°C (2mmHg)
MDL Number MFCD00082658