missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Di-n-hexylacetamide, 98+%

Manufacturer:  Alfa Aesar™ L1080518

 View more versions of this product

Catalog No. AAL1080518



Specifications

Specifications

N,N-Di-n-hexylacetamide
130°C (2mmHg)
16423-51-1
MFCD00082658
n,n-di-n-hexylacetamide, n-dihexyl-acetamid, acetamide, n,n-dihexyl, n,n-di-n-hexylacetamide, 98+%
CCCCCCN(CCCCCC)C(=O)C
227.392
227.39
106°C (223°F)
50g
C14H29NO
1772656
JFUCOPIALLYRGN-UHFFFAOYSA-N
N,N-dihexylacetamide
243972
≥98%

Chemical Identifiers

16423-51-1
227.392
JFUCOPIALLYRGN-UHFFFAOYSA-N
243972
CCCCCCN(CCCCCC)C(=O)C
C14H29NO
MFCD00082658
n,n-di-n-hexylacetamide, n-dihexyl-acetamid, acetamide, n,n-dihexyl, n,n-di-n-hexylacetamide, 98+%
N,N-dihexylacetamide
Documents