Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Di-n-butylethylenediamine, 98%

Chemical Identifiers

Chemical Name or Material N,N-Di-n-butylethylenediamine
CAS 3529-09-7
Molecular Formula C10H24N2
Formula Weight 172.32g/mol
MDL Number MFCD00042827
UN Number UN2735
Beilstein 1738440
Synonym 2-(Di-n-butylamino)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1153709

 
alfa aesar™
L11537-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 109° to 114°C (20mmHg)
Quantity 10g
Flash Point 88°C (190°F)
Chemical Name or Material N,N-Di-n-butylethylenediamine
CAS 3529-09-7
Assay 98%
Molecular Formula C10H24N2
Formula Weight 172.32g/mol
MDL Number MFCD00042827
UN Number UN2735
Beilstein 1738440
Synonym 2-(Di-n-butylamino)ethylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.