Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Di-n-butyl-1,3-propylenediamine, 97%

Chemical Identifiers

Chemical Name or Material N,N-Di-n-butyl-1,3-propylenediamine
CAS 102-83-0
Molecular Formula C11H26N2
Formula Weight 186.34g/mol
MDL Number MFCD00008219
UN Number UN2922
Beilstein 635829
Synonym 3-(Di-n-butylamino)propylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1004709

 
alfa aesar™
L10047-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1004718

 
alfa aesar™
L10047-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point −70°C
Boiling Point 204° to 206°C
Quantity 10g
Flash Point 104°C (219°F)
Odor Amine-like
Chemical Name or Material N,N-Di-n-butyl-1,3-propylenediamine
CAS 102-83-0
Assay 97%
Molecular Formula C11H26N2
Formula Weight 186.34g/mol
MDL Number MFCD00008219
UN Number UN2922
Beilstein 635829
Synonym 3-(Di-n-butylamino)propylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H314-H318-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECS Number TX7175000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H314-H318-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.