missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Di-Boc-1H-pyrazole-1-carboxamidine, 98+%

Manufacturer:  Alfa Aesar™ H5456503

 View more versions of this product

Catalog No. AAH5456503



Description

Description

Applications
N,N′-Di-Boc-1H-pyrazole-1-carboxamidine is used in the stereoselective synthesis of the bicyclic guanidine alkaloid (+)-monanchorin.

Solubility
Slightly soluble in water. Soluble in methanol, and chloroform.

Notes
Store at 4°C.
Specifications

Specifications

N,N'-Di-Boc-1H-pyrazole-1-carboxamidine
1g
C14H22N4O4
1-n,n'-di-boc amidino pyrazole
QFNFDHNZVTWZED-UHFFFAOYSA-N
tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-pyrazol-1-ylmethylidene]carbamate
6383521
≥98%
86°C to 90°C
152120-54-2
MFCD01075122
Slightly soluble in water. Soluble in methanol,and chloroform.
CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1
310.354
310.35

Chemical Identifiers

152120-54-2
310.354
QFNFDHNZVTWZED-UHFFFAOYSA-N
6383521
CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1
C14H22N4O4
MFCD01075122
1-n,n'-di-boc amidino pyrazole
tert-butyl (NZ)-N-[[(2-methylpropan-2-yl)oxycarbonylamino]-pyrazol-1-ylmethylidene]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents