missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Di(1-naphthyl)benzidine, 98%

$29.45 - $345.43

Chemical Identifiers

CAS 152670-41-2
Molecular Formula C32H24N2
Molecular Weight (g/mol) 436.558
MDL Number MFCD09261380
InChI Key SZDXPEWZZGNIBB-UHFFFAOYSA-N
Synonym n,n'-di 1-naphthyl-4,4'-benzidine, n4,n4'-di naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine, n4,n4'-di-naphthalen-1-yl-biphenyl-4,4'-diamine, n4,n4'-di naphthalen-1-yl biphenyl-4,4'-diamine, n,n'-di 1-naphthyl benzidine, 1,1'-biphenyl-4,4'-diamine,n4,n4'-di-1-naphthalenyl, n, n'-dinaphthol-benzidine, n4,n4'-bis naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine, pubchem12682, acmc-1bwou
PubChem CID 18958605
IUPAC Name N-[4-[4-(naphthalen-1-ylamino)phenyl]phenyl]naphthalen-1-amine
SMILES C1=CC=C2C(=C1)C=CC=C2NC3=CC=C(C=C3)C4=CC=C(C=C4)NC5=CC=CC6=CC=CC=C65
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64535MD View Documents Alfa Aesar™
H64535MD
250mg Each for $29.45
 
AAH6453503 View Documents Alfa Aesar™
H6453503
1g Each for $86.99
 
AAH6453506 View Documents Alfa Aesar™
H6453506
5g Each for $345.43
 
Specifications

Chemical Identifiers

152670-41-2
436.558
SZDXPEWZZGNIBB-UHFFFAOYSA-N
18958605
C1=CC=C2C(=C1)C=CC=C2NC3=CC=C(C=C3)C4=CC=C(C=C4)NC5=CC=CC6=CC=CC=C65
C32H24N2
MFCD09261380
n,n'-di 1-naphthyl-4,4'-benzidine, n4,n4'-di naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine, n4,n4'-di-naphthalen-1-yl-biphenyl-4,4'-diamine, n4,n4'-di naphthalen-1-yl biphenyl-4,4'-diamine, n,n'-di 1-naphthyl benzidine, 1,1'-biphenyl-4,4'-diamine,n4,n4'-di-1-naphthalenyl, n, n'-dinaphthol-benzidine, n4,n4'-bis naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine, pubchem12682, acmc-1bwou
N-[4-[4-(naphthalen-1-ylamino)phenyl]phenyl]naphthalen-1-amine

Specifications

250mg
C32H24N2
3484371
SZDXPEWZZGNIBB-UHFFFAOYSA-N
N-[4-[4-(naphthalen-1-ylamino)phenyl]phenyl]naphthalen-1-amine
18958605
Solid
N,N'-Di(1-naphthyl)benzidine
152670-41-2
MFCD09261380
n,n'-di 1-naphthyl-4,4'-benzidine, n4,n4'-di naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine, n4,n4'-di-naphthalen-1-yl-biphenyl-4,4'-diamine, n4,n4'-di naphthalen-1-yl biphenyl-4,4'-diamine, n,n'-di 1-naphthyl benzidine, 1,1'-biphenyl-4,4'-diamine,n4,n4'-di-1-naphthalenyl, n, n'-dinaphthol-benzidine, n4,n4'-bis naphthalen-1-yl-1,1'-biphenyl-4,4'-diamine, pubchem12682, acmc-1bwou
C1=CC=C2C(=C1)C=CC=C2NC3=CC=C(C=C3)C4=CC=C(C=C4)NC5=CC=CC6=CC=CC=C65
436.558
436.56
98%