missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N′-Carbonyldiimidazole, CDI, Ricca Chemical

Protein Crosslinker

Manufacturer:  Ricca Chemical Company RIRMB135015C2

Catalog No. MB135015C2



Description

Description

Specifications

Specifications

N,N′-Carbonyldiimidazole, CDI
White
5g
530-62-1
100
C7H6N4O
Soluble in water
C1=CN(C=N1)C(=O)N2C=CN=C2
162.152
162.15
98.4% min.
Glass Amber Bottle
117°C to 122°C
7.7 @ 50g/L
100
Quantitative NMR
1,1'-carbonylbis-1h-imidazole, 1,1'-carbonyldiimidazole, 1,1-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, carbonyl diimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, diimidazol-1-yl ketone, n,n'-carbonyldiimidazole, n,n-carbonyldiimidazole
PFKFTWBEEFSNDU-UHFFFAOYSA-N
di(imidazol-1-yl)methanone
68263
Solid
Molecular Biology

Chemical Identifiers

530-62-1
162.152
1,1'-carbonylbis-1h-imidazole, 1,1'-carbonyldiimidazole, 1,1-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, carbonyl diimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, diimidazol-1-yl ketone, n,n'-carbonyldiimidazole, n,n-carbonyldiimidazole
di(imidazol-1-yl)methanone
C7H6N4O
PFKFTWBEEFSNDU-UHFFFAOYSA-N
68263
C1=CN(C=N1)C(=O)N2C=CN=C2
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Recommended Storage : Refridgerated

SDS
Documents