Please login to your online account to display your discounted pricing

N-N-Butyldiethanolamine, 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  154360050

 View more versions of this product

Catalog No. AC154360050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Physical Form Powder
Formula Weight 161.24
Quantity 5mL
Color Red-brown to dark brown
Beilstein 04, 285
Water 0.5% Max.
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H19NO2
MDL Number MFCD00002856
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material N-N-Butyldiethanolamine
Packaging Glass bottle
Refractive Index 1.4610 to 1.4640 (20°C, 589nm)
Linear Formula CH3(CH2)3N(CH2CH2OH)2
CAS 102-79-4
Synonym 2, 2';-(Butylimino)diethanol
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N-N-Butyldiethanolamine
CAS 102-79-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C8H19NO2
Linear Formula CH3(CH2)3N(CH2CH2OH)2
Formula Weight 161.24
MDL Number MFCD00002856
Synonym 2, 2';-(Butylimino)diethanol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents