missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-n-Butylbenzenesulfonamide, 98%

$54.78 - $54.78

Chemical Identifiers

CAS 3622-84-2
Formula Weight 213.3
MDL Number MFCD00025024
Chemical Name or Material N-n-Butylbenzenesulfonamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61945A4 SDS Alfa Aesar™
H61945A4
2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6194536 SDS Alfa Aesar™
H6194536
500g Each for $54.78
Specifications Description & Specifications

Chemical Identifiers

3622-84-2
213.3
MFCD00025024
N-n-Butylbenzenesulfonamide

Specifications

1.150g/mL
Colorless
3622-84-2
Liquid
MFCD00025024
98%
314°C
2.5kg
1.525
213.3
N-n-Butylbenzenesulfonamide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DB1283000

TSCA : TSCA