Learn More
N-(n-Butyl)ethylenediamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1343714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-(n-Butyl)ethylenediamine | |
0.836 | |
59°C (138°F) | |
1.443 | |
25 g | |
1732700 | |
n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine | |
CCCCNCCN | |
116.208 | |
116.21 |
19522-69-1 | |
170°C to 171°C | |
C6H16N2 | |
MFCD00025602 | |
UN2734 | |
Air Sensitive | |
DFPGBRPWDZFIPP-UHFFFAOYSA-N | |
N'-butylethane-1,2-diamine | |
519668 | |
97% |
Chemical Identifiers
19522-69-1 | |
116.208 | |
DFPGBRPWDZFIPP-UHFFFAOYSA-N | |
519668 | |
CCCCNCCN |
C6H16N2 | |
MFCD00025602 | |
n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine | |
N'-butylethane-1,2-diamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only