missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-n-Butyl-4-nitrobenzamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 51207-98-8
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.244
MDL Number MFCD00157824
InChI Key ZPYLRHGQFFDNOH-UHFFFAOYSA-N
Synonym benzamide, n-butyl-4-nitro, n-butyl-4-nitro-benzamide, 4-nitro-n-butylbenzamide, n-n-butyl-4-nitrobenzamide, benzamide, n-butyl-p-nitro, n-butyl 4-nitrophenyl carboxamide
PubChem CID 347929
IUPAC Name N-butyl-4-nitrobenzamide
SMILES CCCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60919MD View Documents Alfa Aesar™
H60919MD
250mg Each for $220.53
 
AAH6091903 View Documents Alfa Aesar™
H6091903
1g Each for $703.20
 
Specifications

Chemical Identifiers

51207-98-8
222.244
ZPYLRHGQFFDNOH-UHFFFAOYSA-N
347929
CCCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C11H14N2O3
MFCD00157824
benzamide, n-butyl-4-nitro, n-butyl-4-nitro-benzamide, 4-nitro-n-butylbenzamide, n-n-butyl-4-nitrobenzamide, benzamide, n-butyl-p-nitro, n-butyl 4-nitrophenyl carboxamide
N-butyl-4-nitrobenzamide

Specifications

250mg
C11H14N2O3
benzamide, n-butyl-4-nitro, n-butyl-4-nitro-benzamide, 4-nitro-n-butylbenzamide, n-n-butyl-4-nitrobenzamide, benzamide, n-butyl-p-nitro, n-butyl 4-nitrophenyl carboxamide
CCCCNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
222.244
222.24
N-n-Butyl-4-nitrobenzamide
51207-98-8
MFCD00157824
ZPYLRHGQFFDNOH-UHFFFAOYSA-N
N-butyl-4-nitrobenzamide
347929
97%