missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-n-Butyl-4-chloro-3-nitrobenzamide, 97%

$208.93 - $668.59

Chemical Identifiers

CAS 59320-12-6
Molecular Formula C11H13ClN2O3
Molecular Weight (g/mol) 256.686
MDL Number MFCD01182368
InChI Key YVBGUSUYCUSCBC-UHFFFAOYSA-N
Synonym n-butyl 4-chloro-3-nitrophenyl carboxamide
PubChem CID 1722534
IUPAC Name N-butyl-4-chloro-3-nitrobenzamide
SMILES CCCCNC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59333MD View Documents Alfa Aesar™
H59333MD
250mg Each for $208.93
 
AAH5933303 View Documents Alfa Aesar™
H5933303
1g Each for $668.59
 
Specifications

Chemical Identifiers

59320-12-6
256.686
YVBGUSUYCUSCBC-UHFFFAOYSA-N
1722534
CCCCNC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]
C11H13ClN2O3
MFCD01182368
n-butyl 4-chloro-3-nitrophenyl carboxamide
N-butyl-4-chloro-3-nitrobenzamide

Specifications

250mg
C11H13ClN2O3
1984417
YVBGUSUYCUSCBC-UHFFFAOYSA-N
N-butyl-4-chloro-3-nitrobenzamide
1722534
97%
59320-12-6
MFCD01182368
n-butyl 4-chloro-3-nitrophenyl carboxamide
CCCCNC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]
256.686
256.69
N-n-Butyl-4-chloro-3-nitrobenzamide