Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-Bis(p-toluoyl)hydrazine

$70.07 - $70.07

Chemical Identifiers

Chemical Name or Material N,N'-Bis(p-toluoyl)hydrazine
CAS 1530-73-0
Molecular Formula C16H16N2O4
Formula Weight 300.31
MDL Number MFCD00429172
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1977203

  SDS
alfa aesar™
A1977203
1g Each for $70.07
Description & Specifications

Specifications

Formula Weight 300.31
Chemical Name or Material N,N'-Bis(p-toluoyl)hydrazine
Quantity 1g
Molecular Formula C16H16N2O4
MDL Number MFCD00429172
CAS 1530-73-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.