missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5409206

 View more versions of this product

Catalog No. AAH5409206



Description

Description

N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

N,N'-Bis(benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine
87°C to 91°C
MFCD02683516
n,n'-bis-z-1-guanylpyrazole, n,n'-bis benzyloxycarbonyl-1h-pyrazole-1-carboxamidine, n,n-bisbenzyloxycarbonyl-1-guanylpyrazole, benzyl benzyloxy carbonyl amino 1h-pyrazol-1-yl methylene carbamate, pyrazole z 2, pyrazol z 2, n,n'-bs-z-1-guanylpyrazole, n,n;-bis-z-1-guanylpyrazole, benzyl n-1z-benzyloxy carbonyl imino pyrazol-1-yl methyl carbamate, n,n'-di-cbz-1h-pyrazole-1-carbo
C1=CC=C(C=C1)COC(=O)NC(=NC(=O)OCC2=CC=CC=C2)N3C=CC=N3
378.388
378.38
152120-55-3
C20H18N4O4
5 g
NRBUVVTTYMTSKM-UHFFFAOYSA-N
benzyl (NZ)-N-[phenylmethoxycarbonylamino(pyrazol-1-yl)methylidene]carbamate
6371643
≥98%

Chemical Identifiers

152120-55-3
378.388
NRBUVVTTYMTSKM-UHFFFAOYSA-N
6371643
C1=CC=C(C=C1)COC(=O)NC(=NC(=O)OCC2=CC=CC=C2)N3C=CC=N3
C20H18N4O4
MFCD02683516
n,n'-bis-z-1-guanylpyrazole, n,n'-bis benzyloxycarbonyl-1h-pyrazole-1-carboxamidine, n,n-bisbenzyloxycarbonyl-1-guanylpyrazole, benzyl benzyloxy carbonyl amino 1h-pyrazol-1-yl methylene carbamate, pyrazole z 2, pyrazol z 2, n,n'-bs-z-1-guanylpyrazole, n,n;-bis-z-1-guanylpyrazole, benzyl n-1z-benzyloxy carbonyl imino pyrazol-1-yl methyl carbamate, n,n'-di-cbz-1h-pyrazole-1-carbo
benzyl (NZ)-N-[phenylmethoxycarbonylamino(pyrazol-1-yl)methylidene]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only