Learn More
N,N'-Bis(acryloyl)cystamine, Electrophoresis Reagent, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6689306
Description
BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.
Solubility
Soluble at 50 mg/ml in acetic acid
Notes
Store at -20°C. Incompatible with heat, oxidizing agents.
Specifications
N,N'-Bis(acryloyl)cystamine | |
60984-57-8 | |
White | |
MFCD00036225 | |
n,n'-bis acryloyl cystamine, n,n'-bisacrylylcystamine, 2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis, 2,2'-bisacrylamino diethyldisulfide, bis-acrylylcystamine: bac, n,n'-dithiodi-2,1-ethanediyl bis acrylamide, n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide, n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide, acmc-20ak9p, n,n-bis-acrylylcystamine | |
DJVKJGIZQFBFGS-UHFFFAOYSA-N | |
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide | |
100602 | |
98% |
Electrophoresis Reagent | |
123°C to 124°C | |
C10H16N2O2S2 | |
5 g | |
Soluble at 50mg/mL in acetic acid | |
C=CC(=O)NCCSSCCNC(=O)C=C | |
260.37 | |
260.37 | |
Powder |
Chemical Identifiers
60984-57-8 | |
260.37 | |
DJVKJGIZQFBFGS-UHFFFAOYSA-N | |
100602 | |
C=CC(=O)NCCSSCCNC(=O)C=C |
C10H16N2O2S2 | |
MFCD00036225 | |
n,n'-bis acryloyl cystamine, n,n'-bisacrylylcystamine, 2-propenamide, n,n'-dithiodi-2,1-ethanediyl bis, 2,2'-bisacrylamino diethyldisulfide, bis-acrylylcystamine: bac, n,n'-dithiodi-2,1-ethanediyl bis acrylamide, n-2-2-prop-2-enamido ethyl disulfanyl ethyl prop-2-enamide, n-2-2-prop-2-enoylamino ethyldisulfanyl ethyl prop-2-enamide, acmc-20ak9p, n,n-bis-acrylylcystamine | |
N-[2-[2-(prop-2-enoylamino)ethyldisulfanyl]ethyl]prop-2-enamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 262-546-8
TSCA : No
storageNote1 : −30°C to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only