Please login to your online account to display your discounted pricing

N,N'-Bis(acryloyl)cystamine, 98%, Alfa Aesar™

Used as a reversible cross-linker for polyacrylamide gels

$1,143.69 - $1,143.69

Chemical Identifiers

Chemical Name or Material N,N'-Bis(acryloyl)cystamine
CAS 60984-57-8
Molecular Formula C10H16N2O2S2
Linear Formula (CH2=CHC(O)NHCH2CH2S)2
Formula Weight 260.37
MDL Number MFCD00036225
Synonym BAC
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4413214

 
alfa aesar
4413214
Packaged under Argon 25g Each for $1,143.69
Description & Specifications

Specifications

Melting Point 123° to 124°C
Formula Weight 260.37
Quantity 25g
Chemical Name or Material N,N'-Bis(acryloyl)cystamine
Color White
Physical Form Powder
Assay 98%
Molecular Formula C10H16N2O2S2
Linear Formula (CH2=CHC(O)NHCH2CH2S)2
MDL Number MFCD00036225
CAS 60984-57-8
Synonym BAC
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 −30° to −10°C
Packaging Packaged under Argon
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C