Please login to your online account to display your discounted pricing

N,N-Bis(2-hydroxyethyl)-N'-(α,α-dimethyl-3-isopropenylbenzyl)urea, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  402540250

 View more versions of this product

Catalog No. AC402540250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Crystals
Formula Weight 306.4
Quantity 25g
Color White to light yellow
Melting Point 76° to 80°C
Chemical Name or Material N, N-Bis(2-hydroxyethyl)-N'-(α, α-dimethyl-3-isopropenylbenzyl)urea
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00144975
CAS 111256-33-8
Molecular Formula C17 H26 N2 O3

Specifications

Chemical Name or Material N, N-Bis(2-hydroxyethyl)-N'-(α, α-dimethyl-3-isopropenylbenzyl)urea
CAS 111256-33-8
Molecular Formula C17 H26 N2 O3
Formula Weight 306.4
MDL Number MFCD00144975