Please login to your online account to display your discounted pricing

N,N-Bis(2-hydroxyethyl)-N'-(α,α-dimethyl-3-isopropenylbenzyl)urea, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N, N-Bis(2-hydroxyethyl)-N'-(α, α-dimethyl-3-isopropenylbenzyl)urea
CAS 111256-33-8
Molecular Formula C17 H26 N2 O3
Formula Weight 306.4
MDL Number MFCD00144975
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC402540050

  SDS
thermo fisher scientific
402540050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC402540250

  SDS
thermo fisher scientific
402540250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Crystals
Formula Weight 306.4
Quantity 5g
Color White to light yellow
Melting Point 76° to 80°C
Chemical Name or Material N, N-Bis(2-hydroxyethyl)-N'-(α, α-dimethyl-3-isopropenylbenzyl)urea
Infrared Spectrum Authentic
Molecular Formula C17 H26 N2 O3
CAS 111256-33-8
Packaging Glass bottle
MDL Number MFCD00144975