missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N′-Bis(2-hydroxyethyl)ethylenediamine, TCI America™

$70.28 - $70.28

Chemical Identifiers

CAS 4439-20-7
Molecular Formula C6H16N2O2
Molecular Weight (g/mol) 148.206
MDL Number MFCD00002842
InChI Key GFIWSSUBVYLTRF-UHFFFAOYSA-N
Synonym n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol, n,n'-bis 2-hydroxyethyl ethylene diamine
PubChem CID 78179
IUPAC Name 2-[2-(2-hydroxyethylamino)ethylamino]ethanol
SMILES C(CNCCO)NCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B380725G View Documents TCI America
B380725G
25g Each for $70.28
 
Specifications

Chemical Identifiers

4439-20-7
148.206
GFIWSSUBVYLTRF-UHFFFAOYSA-N
78179
C(CNCCO)NCCO
C6H16N2O2
MFCD00002842
n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol, n,n'-bis 2-hydroxyethyl ethylene diamine
2-[2-(2-hydroxyethylamino)ethylamino]ethanol

Specifications

White-Yellow
25g
C6H16N2O2
3259
GFIWSSUBVYLTRF-UHFFFAOYSA-N
2-[2-(2-hydroxyethylamino)ethylamino]ethanol
78179
Crystal-Powder at 20°C
N,N′-Bis(2-hydroxyethyl)ethylenediamine
99°C
4439-20-7
MFCD00002842
n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol, n,n'-bis 2-hydroxyethyl ethylene diamine
C(CNCCO)NCCO
148.206
148.21
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2409

TSCA : Yes