Learn More
N,N-Bis(2-hydroxyethyl)ethylenediamine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03231206
Description
It is employed as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N,N-Bis(2-hydroxyethyl)ethylenediamine | |
1.069 g/mL | |
Amine-like | |
MFCD00014822 | |
UN2735 | |
Soluble in water. | |
NCCN(CCO)CCO | |
148.21 | |
148.21 |
3197-06-6 | |
174°C to 178°C (3 to 4 mmHg) | |
C6H16N2O2 | |
5 g | |
cyclohexanemethylamine, aminomethylcyclohexane, aminomethyl cyclohexane, cyclohexanemethanamine, 1-cyclohexylmethanamine, hexahydrobenzylamine, 1-cyclohexylmethylamine, 4-aminomethylcyclohexane, c-cyclohexyl-methylamine, chembl1049 | |
CYOIAXUAIXVWMU-UHFFFAOYSA-N | |
2-[(2-aminoethyl)(2-hydroxyethyl)amino]ethan-1-ol | |
76688 | |
Liquid |
Chemical Identifiers
3197-06-6 | |
148.21 | |
CYOIAXUAIXVWMU-UHFFFAOYSA-N |
C6H16N2O2 | |
MFCD00014822 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only