missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N-Bis(2-hydroxyethyl)ethylenediamine

Manufacturer:  Alfa Aesar™ 3231203

 View more versions of this product

Catalog No. AA3231203



Specifications

Specifications

N,N-Bis(2-hydroxyethyl)ethylenediamine
Amine-like
1g
C6H16N2O2
UN2735
CYOIAXUAIXVWMU-UHFFFAOYSA-N
2-[2-aminoethyl(2-hydroxyethyl)amino]ethanol
98535
Liquid
1.069g/mL
174°C to 178°C (3 to 4mmHg)
3197-06-6
MFCD00014822
n,n-bis 2-hydroxyethyl ethylenediamine, 2,2'-2-aminoethyl azanediyl diethanol, ethanol, 2,2'-2-aminoethyl imino bis, 2,2'-2-aminoethyl imino bisethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethan-1-ol, aminoethyldiethanolamine, aminoethylethanolamine solution, 2,2'-2-aminoethyl imino diethanol, acmc-20a9el
C(CN(CCO)CCO)N
148.21
148.21

Chemical Identifiers

3197-06-6
148.21
CYOIAXUAIXVWMU-UHFFFAOYSA-N
98535
C(CN(CCO)CCO)N
C6H16N2O2
MFCD00014822
n,n-bis 2-hydroxyethyl ethylenediamine, 2,2'-2-aminoethyl azanediyl diethanol, ethanol, 2,2'-2-aminoethyl imino bis, 2,2'-2-aminoethyl imino bisethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethan-1-ol, aminoethyldiethanolamine, aminoethylethanolamine solution, 2,2'-2-aminoethyl imino diethanol, acmc-20a9el
2-[2-aminoethyl(2-hydroxyethyl)amino]ethanol
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

SDS Documents