missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-(4-Methyl-1,3-phenylene)bis{4-(2-pyridinyl)-1-piperazinecarboxamide}, 1,000 μg/mL, MilliporeSigma™ Supelco™
Description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Specifications
Specifications
| Packaging | Ampule |
| Concentration | 1000 μg/mL in DMSO (as the derivative) |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |