Learn More
N,N,3,5-Tetramethylaniline, 98%, Thermo Scientific Chemicals
$197.62 - $748.34
Chemical Identifiers
CAS | 4913-13-7 |
---|---|
Molecular Formula | C10H15N |
Molecular Weight (g/mol) | 149.237 |
MDL Number | MFCD00008307 |
InChI Key | NBFRQCOZERNGEX-UHFFFAOYSA-N |
Synonym | benzenamine, n,n,3,5-tetramethyl, n,n-dimethyl-3,5-xylidine, 3,5-xylidine, n,n-dimethyl, 3,5-dimethylphenyl dimethylamine, acmc-209kew, 5-dimethylamino-m-xylene, 3,5,n,n-tetramethylaniline, n,n-dimethyl-sym-m-xylidine, n,n,3,5-tetramethyl aniline, n,n-3,5-tetramethylbenzenamine |
PubChem CID | 78625 |
IUPAC Name | N,N,3,5-tetramethylaniline |
SMILES | CC1=CC(=CC(=C1)N(C)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0926606
|
Thermo Scientific Chemicals
L0926606 |
5 g |
Each for $197.62
|
|
|||||
AAL0926614
|
Thermo Scientific Chemicals
L0926614 |
25 g |
Each for $748.34
|
|
|||||
Description
N,N,3,5-Tetramethylaniline was used in rapid quenching of 1,8-dihydroxyanthraquinone (DHAQ). It was also used as a catalyst in the polymeric synthesis of glycol methacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N,3,5-Tetramethylaniline was used in rapid quenching of 1,8-dihydroxyanthraquinone (DHAQ). It was also used as a catalyst in the polymeric synthesis of glycol methacrylate.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
4913-13-7 | |
149.237 | |
NBFRQCOZERNGEX-UHFFFAOYSA-N | |
78625 | |
CC1=CC(=CC(=C1)N(C)C)C |
C10H15N | |
MFCD00008307 | |
benzenamine, n,n,3,5-tetramethyl, n,n-dimethyl-3,5-xylidine, 3,5-xylidine, n,n-dimethyl, 3,5-dimethylphenyl dimethylamine, acmc-209kew, 5-dimethylamino-m-xylene, 3,5,n,n-tetramethylaniline, n,n-dimethyl-sym-m-xylidine, n,n,3,5-tetramethyl aniline, n,n-3,5-tetramethylbenzenamine | |
N,N,3,5-tetramethylaniline |
Specifications
4913-13-7 | |
226°C to 228°C | |
Recognizable | |
C10H15N | |
UN2810 | |
benzenamine, n,n,3,5-tetramethyl, n,n-dimethyl-3,5-xylidine, 3,5-xylidine, n,n-dimethyl, 3,5-dimethylphenyl dimethylamine, acmc-209kew, 5-dimethylamino-m-xylene, 3,5,n,n-tetramethylaniline, n,n-dimethyl-sym-m-xylidine, n,n,3,5-tetramethyl aniline, n,n-3,5-tetramethylbenzenamine | |
NBFRQCOZERNGEX-UHFFFAOYSA-N | |
N,N,3,5-tetramethylaniline | |
78625 | |
98% |
0.913 | |
5 g | |
1.544 | |
MFCD00008307 | |
1933736 | |
Not miscible or difficult to mix with water. | |
CC1=CC(=CC(=C1)N(C)C)C | |
149.237 | |
149.24 | |
N,N,3,5-Tetramethylaniline |
Safety and Handling
GHS H Statement
H311-H373-H227-H302-H332
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
P210-P235-P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 225-539-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only