missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,2,4,6-Pentamethylaniline, TCI America™

Manufacturer:  TCI America P176725G

 View more versions of this product

Catalog No. P176725G



Specifications

Specifications

N,N,2,4,6-Pentamethylaniline
Yellow
13021-15-3
MFCD00010596
JZBZLRKFJWQZHU-UHFFFAOYSA-N
N,N,2,4,6-pentamethylaniline
139365
Clear Liquid at 20°C
215°C
25g
C11H17N
n,n,2,4,6-pentamethylbenzenamine, 2-dimethylamino mesitylene, n,n-dimethylmesidine, benzenamine,n,n,2,4,6-pentamethyl, 2-dimethylaminomesitylene, dimethylaminomesitylen, pubchem3219, acmc-209bhq, n,n-2,4,6-pentamethylaniline, 2,4,6,n,n-pentamethylbenzenamine
CC1=CC(=C(C(=C1)C)N(C)C)C
163.264
163.26
≥97.0% (GC)

Chemical Identifiers

13021-15-3
163.264
JZBZLRKFJWQZHU-UHFFFAOYSA-N
139365
CC1=CC(=C(C(=C1)C)N(C)C)C
C11H17N
MFCD00010596
n,n,2,4,6-pentamethylbenzenamine, 2-dimethylamino mesitylene, n,n-dimethylmesidine, benzenamine,n,n,2,4,6-pentamethyl, 2-dimethylaminomesitylene, dimethylaminomesitylen, pubchem3219, acmc-209bhq, n,n-2,4,6-pentamethylaniline, 2,4,6,n,n-pentamethylbenzenamine
N,N,2,4,6-pentamethylaniline
Safety and Handling

Safety and Handling

TSCA : No

Documents