missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,2,2-Tetramethyl-1,3-propanediamine, 97%, ACROS Organics™

Chemical Identifiers

CAS 53369-71-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H18N2
Linear Formula (CH3)2NCH2C(CH3)2CH2NH2
Formula Weight 130.23
Synonym N, N-Dimethylneopentanediamine
Chemical Name or Material N,N,2,2-Tetramethyl-1,3-propanediamine
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC210870250 SDS Thermo Fisher Scientific
AC210870250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC210871000 SDS Thermo Fisher Scientific
AC210871000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

53369-71-4
100.0
C7H18N2
130.23
N,N,2,2-Tetramethyl-1,3-propanediamine
96.0
97%
(CH3)2NCH2C(CH3)2CH2NH2
N, N-Dimethylneopentanediamine
≥96.0%

Specifications

0.82g/mL
25g
96.0
Authentic
Liquid
C7H18N2
1.4345 to 1.4365 (20°C, 589nm)
N, N-Dimethylneopentanediamine
≥96.0%
Clear colorless to light yellow
53369-71-4
100.0
Glass bottle
97%
(CH3)2NCH2C(CH3)2CH2NH2
130.23
N,N,2,2-Tetramethyl-1,3-propanediamine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger