Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N''-1,4-Phenylenebis(thiourea), 98+%

$57.57 - $201.91

Chemical Identifiers

Chemical Name or Material N,N''-1,4-Phenylenebis(thiourea)
CAS 1519-70-6
Molecular Formula C8H10N4S2
Formula Weight 226.32
MDL Number MFCD00022169
Beilstein 2114854
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1210206

  SDS
alfa aesar™
L1210206
5g Each for $201.91

AAL1210203

  SDS
alfa aesar™
L1210203
1g Each for $57.57
Description & Specifications

Specifications

Chemical Name or Material N,N''-1,4-Phenylenebis(thiourea)
CAS 1519-70-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number YU1292010
Melting Point 218° to 222°C
Assay ≥98%
Quantity 5g
Molecular Formula C8H10N4S2
Odor Odorless
Formula Weight 226.32
MDL Number MFCD00022169
Beilstein 2114854
Safety and Handling

Warning

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.