missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N''-1,4-Phenylenebis(thiourea), 98+%

Manufacturer:  Alfa Aesar™ L1210203

 View more versions of this product

Catalog No. AAL1210203



Specifications

Specifications

N,N''-1,4-Phenylenebis(thiourea)
218°C to 222°C
1519-70-6
MFCD00022169
1,4-phenylenebis 2-thiourea, urea, 1,1'-p-phenylenebis 2-thio, 1,1'-p-phenylenebis 2-thiourea, thiourea, n,n-1,4-phenylenebis, 4-carbamothioylamino phenylthiourea, 4-carbamothioylamino phenyl thiourea, 1,1'-benzene-1,4-diylbis thiourea, 1,4-phenylenebis, phenylenethiourea, acmc-20ap1t
C1=CC(=CC=C1NC(=S)N)NC(=S)N
226.316
226.32
Odorless
1g
C8H10N4S2
2114854
AMNPXXIGUOKIPP-UHFFFAOYSA-N
[4-(carbamothioylamino)phenyl]thiourea
684351
≥98%

Chemical Identifiers

1519-70-6
226.316
AMNPXXIGUOKIPP-UHFFFAOYSA-N
684351
C1=CC(=CC=C1NC(=S)N)NC(=S)N
C8H10N4S2
MFCD00022169
1,4-phenylenebis 2-thiourea, urea, 1,1'-p-phenylenebis 2-thio, 1,1'-p-phenylenebis 2-thiourea, thiourea, n,n-1,4-phenylenebis, 4-carbamothioylamino phenylthiourea, 4-carbamothioylamino phenyl thiourea, 1,1'-benzene-1,4-diylbis thiourea, 1,4-phenylenebis, phenylenethiourea, acmc-20ap1t
[4-(carbamothioylamino)phenyl]thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YU1292010

Documents