Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-(1,2-Dihydroxyethylene)bisacrylamide, Electrophoresis Reagent, 97%


Quantity 5g

Manufacturer: alfa aesar™  J6689906

 View more versions of this product

Catalog No. AAJ6689906

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.2
Melting Point 141° to 143°C
MDL Number MFCD00008624
Synonym 1,2-Bis(acrylamido)ethylene glycol; DHEBA
Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
Physical Form Powder
Quantity 5g
Beilstein 1941875
CAS 868-63-3
Health Hazard 1 Warning
Name Note Electrophoresis Reagent
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
Assay 97%
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Color White to cream

Specifications

Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
CAS 868-63-3
Name Note Electrophoresis Reagent
Formula Weight 200.2
MDL Number MFCD00008624
Beilstein 1941875
Synonym 1,2-Bis(acrylamido)ethylene glycol; DHEBA

  • A cross-linker for polyacrylamide gels
  • Useful when recovering protein from polyacrylamide gels

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C

SDS & Safety Documents