Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-(1,2-Dihydroxyethylene)bisacrylamide, Electrophoresis Reagent, 97%

Chemical Identifiers

Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
CAS 868-63-3
Name Note Electrophoresis Reagent
Formula Weight 200.2
MDL Number MFCD00008624
Beilstein 1941875
Synonym 1,2-Bis(acrylamido)ethylene glycol; DHEBA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6689906

 
alfa aesar™
J6689906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ6689914

 
alfa aesar™
J6689914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.2
Melting Point 141° to 143°C
Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
Name Note Electrophoresis Reagent
Physical Form Powder
MDL Number MFCD00008624
CAS 868-63-3
Beilstein 1941875
Synonym 1,2-Bis(acrylamido)ethylene glycol; DHEBA
Health Hazard 1 Warning
Quantity 5g
TSCA TSCA
Assay 97%
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
Color White to cream

  • A cross-linker for polyacrylamide gels
  • Useful when recovering protein from polyacrylamide gels

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C