Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-(1,2-Dihydroxyethylene)bisacrylamide, Electrophoresis Reagent, 97%


Quantity 25g

Manufacturer: alfa aesar™  J6689914

 View more versions of this product

Catalog No. AAJ6689914

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.2
Melting Point 141° to 143°C
Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
TSCA TSCA
Physical Form Powder
Assay 97%
MDL Number MFCD00008624
CAS 868-63-3
Beilstein 1941875
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White to cream
Quantity 25g
Name Note Electrophoresis Reagent
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Storage Note 1 2° to 8°C
Synonym 1,2-Bis(acrylamido)ethylene glycol; DHEBA

Specifications

Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
CAS 868-63-3
Name Note Electrophoresis Reagent
Formula Weight 200.2
MDL Number MFCD00008624
Beilstein 1941875
Synonym 1,2-Bis(acrylamido)ethylene glycol; DHEBA

  • A cross-linker for polyacrylamide gels
  • Useful when recovering protein from polyacrylamide gels

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C