Please login to your online account to display your discounted pricing

Alfa Aesar™ N,N'-(1,2-Dihydroxyethylene)bisacrylamide, 97%

$456.69 - $456.69

Chemical Identifiers

Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
CAS 868-63-3
Molecular Formula C8H12N2O4
Formula Weight 200.20g/mol
MDL Number MFCD00008624
Beilstein 1941875
Synonym 1,2-Bis(acrylamido)ethylene glycol; DHEBA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1921114

 
alfa aesar™
L19211-14
25g Each for $456.69
Description & Specifications

Specifications

Quantity 25g
Melting Point 141° to 143°C
Quantity 25g
Chemical Name or Material N,N'-(1,2-Dihydroxyethylene)bisacrylamide
CAS 868-63-3
Assay 97%
Molecular Formula C8H12N2O4
Formula Weight 200.20g/mol
MDL Number MFCD00008624
Beilstein 1941875
Synonym 1,2-Bis(acrylamido)ethylene glycol; DHEBA
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
TSCA TSCA

Applications
A cross-linking reagent for the recovery of separated biomoleculesN,N′-(1,2-Dihydroxyethylene)bisacrylamide acts as a cross-linking reagent used for the recovery of separated biomolecules such as proteins from polyacrylamide gels. It is also employed in the formation of gradient gels.

Solubility
Soluble in water and sodium hydroxide.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C