missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-MPPP Hydrochloride, Tocris

Selective κ agonist

$205.70 - $862.95

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C21H26N2O·HCl
Formula Weight 358.91
Synonym N-Methyl-N-[(1S)-1-phenyl-2-(1-pyrrolidinyl)ethyl]phenylacetamide hydrochloride
Chemical Name or Material N-MPPP Hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
078350 R & D Systems
078350
50mg Each for $862.95
078310 R & D Systems
078310
10mg Each for $205.70
Specifications Description & Specifications

Chemical Identifiers

>98%
358.91
N-MPPP Hydrochloride
C21H26N2O·HCl
N-Methyl-N-[(1S)-1-phenyl-2-(1-pyrrolidinyl)ethyl]phenylacetamide hydrochloride

Specifications

50mg
>98%
358.91
N-MPPP Hydrochloride
207452-97-9
C21H26N2O·HCl
N-Methyl-N-[(1S)-1-phenyl-2-(1-pyrrolidinyl)ethyl]phenylacetamide hydrochloride

Safety and Handling

Recommended Storage : Store at Room Temperature