missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nω-Methyltryptamine 99%, ACROS Organics™

Manufacturer:  Acros Organics 151751000

 View more versions of this product

Catalog No. AC151751000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-ω-Methyltryptamine
87.0°C to 91.0°C
99%
98.5
Authentic
99%
C11H14N2
MFCD00005657
NCIKQJBVUNUXLW-UHFFFAOYSA-N
2-(1H-indol-3-yl)-N-methylethanamine
6088
174.24
White
100mg
61-49-4
100.0
Glass bottle
99%
1% max. (K.F.)
3-(2-Methylaminoethyl)indole
CNCCC1=CNC2=CC=CC=C21
174.247
CHEBI:28136
Shiny Crystalline Powder or Flakes

Chemical Identifiers

61-49-4
174.247
NCIKQJBVUNUXLW-UHFFFAOYSA-N
6088
C11H14N2
MFCD00005657
3-(2-Methylaminoethyl)indole
CHEBI:28136
Safety and Handling

Safety and Handling

EINECSNumber : 200-507-9

RTECSNumber : NM0450000

SDS Documents