Please login to your online account to display your discounted pricing

Nω-Methyltryptamine 99%, ACROS Organics™

$425.58 - $425.58

Chemical Identifiers

Chemical Name or Material Nomega-Methyltryptamine
Name Note 99%
CAS 61-49-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H14N2
Formula Weight 174.24
MDL Number MFCD00005657
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC151750010

  SDS
thermo fisher scientific
151750010
Glass bottle 1g Each for $425.58

AC151751000

  SDS
thermo fisher scientific
151751000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Shiny Crystalline Powder or Flakes
Formula Weight 174.24
Quantity 1g
Color White to off-white
RTECS Number NM0450000
EINECS Number 200-507-9
Melting Point 87° to 91°C
CAS Max % 100.0
Synonym 3-(2-Methylaminoethyl)indole
Infrared Spectrum Authentic
CAS Min % 98.5
Water 1% max. (K.F.)
Packaging Glass bottle
Assay Percent Range 99%
Chemical Name or Material Nomega-Methyltryptamine
Name Note 99%
Molecular Formula C11H14N2
CAS 61-49-4
MDL Number MFCD00005657