missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyltrimethylacetamide, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0891606

 View more versions of this product

Catalog No. AAL0891606


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Methyltrimethylacetamide
89°C to 93°C
C6H13NO
MFCD00043643
1744142
QMKKJBRRKIKWFK-UHFFFAOYSA-N
N,2,2-trimethylpropanamide
138844
96%
6830-83-7
105°C to 110°C (15 mmHg)
(CH3)3CCONHCH3
5 g
n-methylpivalamide, n-methyltrimethylacetamide, 2,2,n-trimethylpropanamide, n,2,2-trimethylpropionamide, n-methyl pivalamide, 2,2-dimethyl-n-methylpropanamide, bae, n-methyl-pivalamide, propanamide,n,2,2-trimethyl
CC(C)(C)C(=O)NC
115.176
115.18

Chemical Identifiers

6830-83-7
115.176
QMKKJBRRKIKWFK-UHFFFAOYSA-N
138844
CC(C)(C)C(=O)NC
C6H13NO
MFCD00043643
n-methylpivalamide, n-methyltrimethylacetamide, 2,2,n-trimethylpropanamide, n,2,2-trimethylpropionamide, n-methyl pivalamide, 2,2-dimethyl-n-methylpropanamide, bae, n-methyl-pivalamide, propanamide,n,2,2-trimethyl
N,2,2-trimethylpropanamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only