missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methylthiourea 98.0+%, TCI America™
Supplier: TCI America M0619100G
Specifications
N-Methylthiourea | |
120°C | |
7 (100g/L H2O soln. 20°C) | |
MFCD00004938 | |
2811 | |
KQJQICVXLJTWQD-UHFFFAOYSA-N | |
methylthiourea | |
2723704 | |
≥98.0% (T) |
598-52-7 | |
White-Yellow | |
C2H6N2S | |
100 g | |
n-methylthiourea, 1-methylthiourea, 1-methyl-2-thiourea, methylthiocarbamide, thiourea, methyl, monomethylthiourea, 1-methylthiocarbamide, methyl thiourea, n-methylthiocarbamide, urea, 1-methyl-2-thio | |
CNC(=S)N | |
90.144 | |
90.14 | |
Crystalline Powder |
Chemical Identifiers
598-52-7 | |
90.144 | |
KQJQICVXLJTWQD-UHFFFAOYSA-N | |
2723704 | |
CNC(=S)N |
C2H6N2S | |
MFCD00004938 | |
n-methylthiourea, 1-methylthiourea, 1-methyl-2-thiourea, methylthiocarbamide, thiourea, methyl, monomethylthiourea, 1-methylthiocarbamide, methyl thiourea, n-methylthiocarbamide, urea, 1-methyl-2-thio | |
methylthiourea |
Safety and Handling
RTECSNumber : YT9000000
TSCA : No