Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylsulfanilic acid, 98+%

$84.97 - $281.69

Chemical Identifiers

Chemical Name or Material N-Methylsulfanilic acid
CAS 24447-99-2
Molecular Formula C7H9NO3S
Formula Weight 187.21
MDL Number MFCD00014732
Beilstein 2094129
Synonym 4-Methylaminobenzenesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0101606

 
alfa aesar™
L0101606
5g Each for $84.97

AAL0101614

 
alfa aesar™
L0101614
25g Each for $281.69
Description & Specifications

Specifications

Formula Weight 187.21
Chemical Name or Material N-Methylsulfanilic acid
Assay ≥98%
CAS 24447-99-2
Synonym 4-Methylaminobenzenesulfonic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈260°C
Quantity 5g
Molecular Formula C7H9NO3S
MDL Number MFCD00014732
Beilstein 2094129
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.