Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methylsuccinimide, 98%

$50.65 - $558.76

Chemical Identifiers

Chemical Name or Material N-Methylsuccinimide
CAS 1121-07-9
Molecular Formula C5H7NO2
Formula Weight 113.13
MDL Number MFCD00005517
Beilstein 110534
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1295606

 
alfa aesar™
A1295606
5g Each for $50.65

AAA1295622

 
alfa aesar™
A1295622
100g Each for $558.76
Description & Specifications

Specifications

Formula Weight 113.13
Chemical Name or Material N-Methylsuccinimide
Melting Point 63° to 68°C
Boiling Point 234° to 235°C
Assay 98%
Quantity 5g
Molecular Formula C5H7NO2
MDL Number MFCD00005517
CAS 1121-07-9
Beilstein 110534
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UY1028650
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.