Please login to your online account to display your discounted pricing

N-Methylpyrrole-3-boronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Methylpyrrole-3-boronic acid pinacol ester
CAS 953040-54-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥88.0% (GC)
Assay Percent Range 97%
Molecular Formula C11H18BNO2
Formula Weight 207.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC441000010

  SDS
thermo fisher scientific
AC441000010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441000050

  SDS
thermo fisher scientific
AC441000050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.08
Physical Form Powder
Assay ≥88.0% (GC)
Quantity 1g
Color White to off-white
Melting Point 43° to 52°C
CAS Max % 100.0
CAS Min % 96.0
Chemical Name or Material N-Methylpyrrole-3-boronic acid pinacol ester
Molecular Formula C11H18BNO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
CAS 953040-54-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.