Please login to your online account to display your discounted pricing

N-Methylpyrrole-2-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Methylpyrrole-2-carboxylic acid
Name Note 97%
CAS 6973-60-0
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C6H7NO2
Formula Weight 125.13g/mol
MDL Number MFCD00003088
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166950050

  SDS
fisher scientific acros
166950050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 131° to 138°C
Color Beige to brown-gray
Quantity 5g
Chemical Name or Material N-Methylpyrrole-2-carboxylic acid
Name Note 97%
CAS 6973-60-0
CAS Min % 96.0
CAS Max % 100.0
Physical Form Beige to brown-gray powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C6H7NO2
Formula Weight 125.13g/mol
MDL Number MFCD00003088
Beilstein 22, 24
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 230-217-8
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.