missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylpyrrole-2-carboxaldehyde, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC156931000

 View more versions of this product

Catalog No. AC156931000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Methylpyrrole-2-carboxaldehyde
Authentic
1.5580 to 1.5630 (20°C, 589nm)
100g
97.5
98%
MFCD00003087
1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
CN1C=CC=C1C=O
109.128
109.13
≥97.5% (GC)
1.0100g/mL
Glass bottle
Undesignated
1192-58-1
100.0
C6H7NO
21, I, 279
OUKQTRFCDKSEPL-UHFFFAOYSA-N
1-methylpyrrole-2-carbaldehyde
14504
Liquid

Chemical Identifiers

1192-58-1
109.128
OUKQTRFCDKSEPL-UHFFFAOYSA-N
14504
CN1C=CC=C1C=O
C6H7NO
MFCD00003087
1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
1-methylpyrrole-2-carbaldehyde
Safety and Handling

Safety and Handling

Warning

SDS Documents