missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylpropionamide, TCI America™

Manufacturer:  TCI America M048825ML

 View more versions of this product

Catalog No. M048825ML



Specifications

Specifications

N-Methylpropionamide
Colorless
1187-58-2
MFCD00009300
QJQAMHYHNCADNR-UHFFFAOYSA-N
N-methylpropanamide
14470
Clear Liquid at 20°C
147°C
25mL
C4H9NO
n-methylpropionamide, propanamide, n-methyl, propionamide, n-methyl, n-methylpropionic acid amide, methyl propionic acid amide, n-methylpropionsaureamid, unii-2tsz8hw6dq, n-methylpropionsaureamid german, 2tsz8hw6dq, n-methyl propionamide
CCC(=O)NC
87.122
87.12
≥99.0% (GC)

Chemical Identifiers

1187-58-2
87.122
QJQAMHYHNCADNR-UHFFFAOYSA-N
14470
CCC(=O)NC
C4H9NO
MFCD00009300
n-methylpropionamide, propanamide, n-methyl, propionamide, n-methyl, n-methylpropionic acid amide, methyl propionic acid amide, n-methylpropionsaureamid, unii-2tsz8hw6dq, n-methylpropionsaureamid german, 2tsz8hw6dq, n-methyl propionamide
N-methylpropanamide
Safety and Handling

Safety and Handling

RTECSNumber : UE4515000

TSCA : Yes

Documents