missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylpropionamide, TCI America™

$100.83 - $294.86

Chemical Identifiers

CAS 1187-58-2
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00009300
InChI Key QJQAMHYHNCADNR-UHFFFAOYSA-N
Synonym n-methylpropionamide, propanamide, n-methyl, propionamide, n-methyl, n-methylpropionic acid amide, methyl propionic acid amide, n-methylpropionsaureamid, unii-2tsz8hw6dq, n-methylpropionsaureamid german, 2tsz8hw6dq, n-methyl propionamide
PubChem CID 14470
IUPAC Name N-methylpropanamide
SMILES CCC(=O)NC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M048825ML View Documents TCI America
M048825ML
25mL Each for $294.86
 
M04885ML View Documents TCI America
M04885ML
5mL Each for $100.83
 
Specifications

Chemical Identifiers

1187-58-2
87.122
QJQAMHYHNCADNR-UHFFFAOYSA-N
14470
CCC(=O)NC
C4H9NO
MFCD00009300
n-methylpropionamide, propanamide, n-methyl, propionamide, n-methyl, n-methylpropionic acid amide, methyl propionic acid amide, n-methylpropionsaureamid, unii-2tsz8hw6dq, n-methylpropionsaureamid german, 2tsz8hw6dq, n-methyl propionamide
N-methylpropanamide

Specifications

147°C
25mL
C4H9NO
n-methylpropionamide, propanamide, n-methyl, propionamide, n-methyl, n-methylpropionic acid amide, methyl propionic acid amide, n-methylpropionsaureamid, unii-2tsz8hw6dq, n-methylpropionsaureamid german, 2tsz8hw6dq, n-methyl propionamide
CCC(=O)NC
87.122
87.12
≥99.0% (GC)
Colorless
1187-58-2
MFCD00009300
QJQAMHYHNCADNR-UHFFFAOYSA-N
N-methylpropanamide
14470
Clear Liquid at 20°C
N-Methylpropionamide
Safety and Handling

Safety and Handling

RTECSNumber : UE4515000

TSCA : Yes