missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methylpropionamide 99.0+%, TCI America™
Supplier: TCI America M04885ML
Specifications
N-Methylpropionamide | |
Colorless | |
C4H9NO | |
5 mL | |
QJQAMHYHNCADNR-UHFFFAOYSA-N | |
N-methylpropanamide | |
14470 | |
≥99.0% (GC) |
1187-58-2 | |
147°C | |
MFCD00009300 | |
n-methylpropionamide, propanamide, n-methyl, propionamide, n-methyl, n-methylpropionic acid amide, methyl propionic acid amide, n-methylpropionsaureamid, unii-2tsz8hw6dq, n-methylpropionsaureamid german, 2tsz8hw6dq, n-methyl propionamide | |
CCC(=O)NC | |
87.122 | |
87.12 | |
Liquid |
Chemical Identifiers
1187-58-2 | |
87.122 | |
QJQAMHYHNCADNR-UHFFFAOYSA-N | |
14470 | |
CCC(=O)NC |
C4H9NO | |
MFCD00009300 | |
n-methylpropionamide, propanamide, n-methyl, propionamide, n-methyl, n-methylpropionic acid amide, methyl propionic acid amide, n-methylpropionsaureamid, unii-2tsz8hw6dq, n-methylpropionsaureamid german, 2tsz8hw6dq, n-methyl propionamide | |
N-methylpropanamide |
Safety and Handling
RTECSNumber : UE4515000
TSCA : Yes