missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylpropargylamine, 98+%, ACROS Organics™

Manufacturer:  Acros Organics AC165540050

 View more versions of this product

Catalog No. AC165540050



Specifications

Specifications

N-Methylpropargylamine
Glass bottle
35161-71-8
100.0
C4H7N
MFCD00008573
HQFYIDOMCULPIW-UHFFFAOYSA-N
N-methylprop-2-yn-1-amine
96160
0.83g/mL
5g
98.0
98+%
HC2CCH2NHCH3
n-methylpropargylamine, 2-propyn-1-amine, n-methyl, methyl-prop-2-ynyl-amine, 3-methylamino-1-propyne, n-methylpropyn-2-ylamine, n-methyl-n-prop-2-ynylamine, n-methyl-2-propyn-1-amine, n-methyl-n-propargylamine, methyl prop-2-yn-1-yl amine, 2-propyn-1-amine,n-methyl-9ci
CNCC#C
69.107
69.11

Chemical Identifiers

35161-71-8
69.107
HQFYIDOMCULPIW-UHFFFAOYSA-N
96160
CNCC#C
C4H7N
MFCD00008573
n-methylpropargylamine, 2-propyn-1-amine, n-methyl, methyl-prop-2-ynyl-amine, 3-methylamino-1-propyne, n-methylpropyn-2-ylamine, n-methyl-n-prop-2-ynylamine, n-methyl-2-propyn-1-amine, n-methyl-n-propargylamine, methyl prop-2-yn-1-yl amine, 2-propyn-1-amine,n-methyl-9ci
N-methylprop-2-yn-1-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Documents