Please login to your online account to display your discounted pricing

N-Methylphthalimide, 98%, Acros Organics


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC305101000

 View more versions of this product

Catalog No. AC305101000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161.16
Quantity 100g
Chemical Name or Material N-Methylphthalimide
CAS 550-44-7
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C9H7NO2

Specifications

Chemical Name or Material N-Methylphthalimide
Assay Percent Range 98%
Molecular Formula C9H7NO2
Formula Weight 161.16