Please login to your online account to display your discounted pricing

n-Methyloctylamine, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC325621000

 View more versions of this product

Catalog No. AC325621000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 143.27
Assay ≥97.5% (Total base) ≥97.5% (GC)
Physical Form Liquid
Quantity 100g
Color Clear colorless to yellow
Density 0.79g/mL
CAS Max % 100.0
Linear Formula CH3(CH)7NHCH3
Chemical Name or Material n-Methyloctylamine
Molecular Formula C9H21N
Refractive Index 1.4260 to 1.4290 (20°C, 589nm)
CAS 2439-54-5
Assay Percent Range 98%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00048927
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material n-Methyloctylamine
CAS 2439-54-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (Total base) ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C9H21N
Linear Formula CH3(CH)7NHCH3
Formula Weight 143.27
MDL Number MFCD00048927
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents