Learn More
N-Methylmorpholine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 127155000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Methylmorpholine | |
109-02-4 | |
100.0 | |
0.9200g/mL | |
14°C | |
98.5% min. (GC) | |
C5H11NO | |
MFCD00006175 | |
27,6 | |
0.92 | |
n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin | |
SJRJJKPEHAURKC-UHFFFAOYSA-N | |
4-methylmorpholine | |
7972 | |
101.15 | |
Liquid |
99% | |
98.5 | |
-65.0°C | |
115.0°C to 116.0°C (750.0 mmHg) | |
Authentic | |
Glass Bottle | |
1.4339 to 1.4359 | |
500 g | |
01,690; 02,278 | |
15,6362 | |
Solubility in water: >500g/L (20°C). Other solubilities: miscible with alcohol,ether and benzene | |
CN1CCOCC1 | |
101.15 | |
0.91 mPa.s (20°C) | |
99% |
Chemical Identifiers
109-02-4 | |
101.15 | |
SJRJJKPEHAURKC-UHFFFAOYSA-N | |
7972 | |
CN1CCOCC1 |
C5H11NO | |
MFCD00006175 | |
n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin | |
4-methylmorpholine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
Ground/Bond container and receiving equipment.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 203-640-0
RTECSNumber : QE5775000
TSCA : TSCA
RUO – Research Use Only